Learn More
Piperonylonitrile, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 4421-09-4 |
|---|---|
| Molecular Formula | C8H5NO2 |
| Molecular Weight (g/mol) | 147.13 |
| MDL Number | MFCD00005820 |
| InChI Key | PKRWWZCDLJSJIF-UHFFFAOYSA-N |
| Synonym | piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358 |
| PubChem CID | 78136 |
| IUPAC Name | 1,3-benzodioxole-5-carbonitrile |
| SMILES | C1OC2=C(O1)C=C(C=C2)C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC151880050
|
Thermo Scientific Chemicals
151880050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 4421-09-4 | |
| 147.13 | |
| PKRWWZCDLJSJIF-UHFFFAOYSA-N | |
| 78136 | |
| C1OC2=C(O1)C=C(C=C2)C#N |
| C8H5NO2 | |
| MFCD00005820 | |
| piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358 | |
| 1,3-benzodioxole-5-carbonitrile |
Specifications
| 4421-09-4 | |
| 97% | |
| C8H5NO2 | |
| 5g | |
| piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358 | |
| C1OC2=C(O1)C=C(C=C2)C#N | |
| 147.13 | |
| 147.13 | |
| Piperonylonitrile, 97% |
| Authentic | |
| Glass bottle | |
| MFCD00005820 | |
| 19, 270 | |
| PKRWWZCDLJSJIF-UHFFFAOYSA-N | |
| 1,3-benzodioxole-5-carbonitrile | |
| 78136 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 224-590-6
RTECSNumber : TO2645000
RUO â Research Use Only