Pipette Supplies Inc PIPTMN/RNN CLASSIC CAL TOOL
Supplier: Pipette Supplies Inc 27777
Pipetman, Rainin Classic Calibration Tool. 5 prong spanner wrench designed for the Gilson Pipetman, Gilson Pipetman G, Gilson Pipetman Neo and the Rainin Classic. Improved design: Long handle to make it easy to hold. This tool is for Pipetman models CxxxxxA and higher, meaning the Pipetman with 6 holes in the top of the black thumb wheel. Instructions included. Note: See item # RA CT-2 for Rainin's Pipet-Lite/Pipet-Plus calibration tool.