missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ 4-Pipette Kit

Gilson™ PIPETMAN™ 4-Pipette Kit features a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids.

Supplier:  Gilson™ F167360

Catalog No. F167360G


Explore available promotions

Description

Description

  • New piston assembly significantly reduces the force needed to aspirate and dispense liquids
  • Come with a durable stainless steel tip ejector
  • Includes three racks of PIPETMAN™ DIAMOND Tips
  • Includes four PIPETMAN™ Pipettes: P2, P20, P200, and P1000G
  • Includes four Single™ Pipette Holders
Specifications

Specifications

Declaration of Conformity
Polypropylene
1
Pipette Kit
14.37 in.
4.72 in.
PIPETMAN™ Diamond Tips
Pipetman™ G
Pipetman™ G
3 years
12.01 in.
4.37 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions