missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ L Pipette Starter Kits

PIPETMAN™ L starter kits provide pipettes covering volume transfers from 0.2 to 1000 μL, which will tackle most laboratory needs. The PIPETMAN™ L pipettes improve on the legendary PIPETMAN™ Classic with a newly designed body for enhanced comfort and a state-of-the-art volume-locking mechanism for precision.

$2027.76 - $2454.50

Specifications

Certifications/Compliance ISO 8655
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Blue
For Use With Manual Liquid Handling
View More Specs

Products 2
Catalog Number Mfr. No. Includes Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Volume (Metric) Price Quantity  
F167350G
View Documents Promotion Details
Gilson™
F167350
PIPETMAN™ L pipettes: P20L, P200L, and P1000L & PIPETMAN™ DIAMOND tips: D200 and D1000 2 to 20 μL (P20L), 20 to 200 μL (P200L), 100 to 1000 μL (P1000L)
Each for $2,027.76
 
F167370G
View Documents Promotion Details
Gilson™
F167370
PIPETMAN™ L pipettes: P2L, P20L, P200L and P1000L & PIPETMAN™ DIAMOND tips: DL10, D200 and D1000 0.2 to 2 μL (P2L), 2 to 20 μL (P20L), 20 to 200 μL (P200L), 100 to 1000 μL (P1000L)
Each for $2,454.50
 
Description

Description

Convenient Use

Lightweight and balanced, PIPETMAN™ L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue.

Right-handed or left-handed? No problem. Adjust the location of the tip ejector to the most comfortable position for you.

Secure Pipetting

Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision.

PIPETMAN™ L variable volume models are all fully autoclavable without disassembly for maximum convenience and protection from cross-contamination.

GLP Compliance

PIPETMAN™ L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.

Everything You Need to Start

Each economically bundled PIPETMAN™ L 3-pipette starter kit (F167350) includes:

  • Three PIPETMAN™ L pipettes: P20L, P200L, and P1000L equipped with plastic tip ejectors
  • Two racks of PIPETMAN™ DIAMOND tips (models: D200 and D1000)
  • Three SINGLE™ pipette holders
  • The Gilson Guide to pipetting
  • The Two-minute inspection poster

Each economically bundled PIPETMAN™ L 4-pipette starter kit (F167370) includes:

  • Four PIPETMAN™ L pipettes: P2L, P20L, P200L and P1000L equipped with stainless-steel tip ejectors
  • Three racks of PIPETMAN™ DIAMOND tips (models: DL10, D200 and D1000)
  • Four SINGLE™ pipette holders
  • The Gilson™ Guide to Pipetting
  • The Two-minute Inspection Poster
Specifications

Specifications

ISO 8655
Autoclavable
Manual Liquid Handling
Pipetman L
Non-sterile
No
Blue
1
No
Pipette Kit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'