Fisherbrand™ Pipette-Specific Tips
Tips for specific applications
Supplier:  Fisherbrand™ 135RAFIS
 View more versions of this product
Catalog No. 21-240-11
Explore available promotions
Description
Description
These tips are specifically designed to fit certain pipettes and are non-sterile for laboratory use.
  • Tips for specific applications
  • Designed to fit distinctive pipettes
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Specifications
1 to 200 μL
Natural
6.35cm
Specialty
Non-sterile
2.5 in.
Stack Rack
Oxford P-3000, Oxford P-7000, Helena