Fisherbrand™ Pipette-Specific Tips
Tips for specific applications
 View more versions of this product
Catalog No. 21-375D
Explore available promotions
Description
Description
These tips are specifically designed to fit certain pipettes and are non-sterile for laboratory use.
  • Tips for specific applications
  • Designed to fit distinctive pipettes
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Specifications
1 to 200 μL
Yellow
4.78cm
Corning Lambda, Eppendorf Reference, Thermo Scientific Finnpipette, Gilson Pipetman, Labpette Digital, Eppendorf Research, Mettler Toledo VoluMate, Nichiryo, Labpette Digital, Gilson Pipetman, Nichiryo, Oxford Benchmate, Gilson Pipetman
Non-sterile
1.88 in.
Bulk