Fisherbrand™ Pipette-Specific Tips
Tips for specific applications
 View more versions of this product
Catalog No. 21-375E
Explore available promotions
Description
Description
These tips are specifically designed to fit certain pipettes and are non-sterile for laboratory use.
  • Tips for specific applications
  • Designed to fit distinctive pipettes
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Specifications
100 to 1000 μL
Blue
7.1cm
Eppendorf
Non-sterile
2.8 in.
Bulk