missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pirfenidone 98.0+%, TCI America™

Supplier:  TCI America P18711G

 View more versions of this product

Catalog No. P18711G


Explore available promotions

Description

Description

Specifications

Specifications

Pirfenidone
109°C
MFCD00866047
pirfenidone, 5-methyl-1-phenylpyridin-2 1h-one, esbriet, deskar, pirespa, amr-69, 5-methyl-1-phenyl-2-1h-pyridone, pirfenidonum, pirfenidona, amr 69
CC1=CN(C(=O)C=C1)C2=CC=CC=C2
185.226
CHEBI:32016
≥98.0% (GC)
53179-13-8
C12H11NO
1 g
ISWRGOKTTBVCFA-UHFFFAOYSA-N
5-methyl-1-phenylpyridin-2-one
40632
185.23
Crystalline Powder

Chemical Identifiers

53179-13-8
185.226
ISWRGOKTTBVCFA-UHFFFAOYSA-N
40632
5-methyl-1-phenylpyridin-2-one
C12H11NO
MFCD00866047
pirfenidone, 5-methyl-1-phenylpyridin-2 1h-one, esbriet, deskar, pirespa, amr-69, 5-methyl-1-phenyl-2-1h-pyridone, pirfenidonum, pirfenidona, amr 69
CHEBI:32016
CC1=CN(C(=O)C=C1)C2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UV1148200

TSCA : No

SDS
Documents

Documents

Promotions

Promotions