missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pivaloyl hydrazide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 377750050

 View more versions of this product

Catalog No. AC377750050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pivaloyl hydrazide
65°C to 67°C
95°C to 110°C (1.0 mbar)
98% min. (GC)
C5H12N2O
MFCD01310839
2,2-dimethylpropionic acid hydrazide, pivalic acid hydrazide, pivalohydrazide, 2,2-dimethylpropanohydrazide, pivaloylhydrazine, 2,2-dimethyl-propionic acid hydrazide, propanoic acid, 2,2-dimethyl-, hydrazide, pivalic acid, hydrazide, 2,2-dimethylpropionicacidhydrazide, trimethylacethydrazide
OARJXUPBZNUYBG-UHFFFAOYSA-N
2,2-dimethylpropanehydrazide
545147
98+%
42826-42-6
White
Authentic
Glass bottle
(CH3)3CC(O)NHNH2
5 g
Solubility in water: .. Other solubilities: soluble in alcohols
CC(C)(C)C(=O)NN
116.16
116.16
Crystals

Chemical Identifiers

42826-42-6
116.16
OARJXUPBZNUYBG-UHFFFAOYSA-N
545147
CC(C)(C)C(=O)NN
C5H12N2O
MFCD01310839
2,2-dimethylpropionic acid hydrazide, pivalic acid hydrazide, pivalohydrazide, 2,2-dimethylpropanohydrazide, pivaloylhydrazine, 2,2-dimethyl-propionic acid hydrazide, propanoic acid, 2,2-dimethyl-, hydrazide, pivalic acid, hydrazide, 2,2-dimethylpropionicacidhydrazide, trimethylacethydrazide
2,2-dimethylpropanehydrazide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only