Plas Labs™ Broome-Style Rodent Restrainers
Ideal for injections or blood sampling
Supplier:  Plas Labs™ 551BSRR
 View more versions of this product
Catalog No. 01-288-32A
Explore available promotions
Description
Description
  • Offer easy access to rodent
  • Prevent rodent from turning
  • Provide safety to the animal handler
  • Manufactured from clear materials to provide complete visibility of animal
Specifications
Specifications
Clear
Rodent Restrainer
Chemically sterilized or washed at 180°F
3.75 in.
1 in.
15 to 30 g
Injections or Blood Sampling
Noryl™ Plastic
Broome
9.52 cm
2.54 cm