Plas Labs™ Model 830 Compact Glove Box
For safe containment of hazardous materials when being weighed
Supplier:  Plas Labs™ 830ABCEXP
 View more versions of this product
Catalog No. 11-389-6
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: White ambidextrous Hypalon™ gloves for superior chemical and U.V. resistance; Manual.
Description
Description
Work chamber accommodates analytical and toploading balances, bench scales and moisture analyzers in a clean, safe, portable and draft-free atmosphere.
  • Top section is removable for easy installation of equipment
  • Four purging gas valves (two on the transfer chamber and two on the main chamber)
  • White leveling tray for easy transferring of liquids
  • All clamps stainless steel and completely adjustable
  • Pressure relief valve minimizes glove fight back
  • HEPA filter on relief valve prevents escape of particulates and organic vapors
  • HEPA and organic vapor canister filters (replaceable)
  • Roomy transfer chamber is 12L x 11 in. I.D.
Certifications
Included hospital-grade multiple electrical outlet strip is UL listed, CSA approved.
Specifications
Specifications
Glove Box
61 cm
31 in.
23 in.
43 in.
24 in.
23 x 43 x 31 in. (61 x 110 x 79 cm)
79 cm
61 cm
110 cm