missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Plastic .6 gal OH pail White
Plastic .6 gal open head pail with screw lid. UN rated 1H2/Y4/S White. HDPE. 7.06 in. dia x 7.39 in.H. Weight is .58 lbs.
Supplier: Youngstown Barrel & Drum 2543NG
Description
Store or ship smaller quantities of solids.Specifications
Open Head | |
Open Head | |
White | |
HDPE | |
Round | |
18.415 cm | |
15.72 cm | |
2.63 cm |
1H2/Y4/S | |
0.6 gal. | |
PACKAGING FOR HAZARDOUS SOLIDS | |
Screw Lid | |
7.25 in. | |
6.19 in. | |
0.58 lb. |