missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives

Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes

Supplier:  Innovative Med Tech 102100

 View more versions of this product

Catalog No. 22-757-143


Explore available promotions

Description

Description

  • Made from 100% plastic
  • Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening

Compliance

Meets FDA, OSHA, and CDC mandates

Specifications

Specifications

Capillary Tube
Plastic
100 mm
2.35 mm
Balanced Li-Heparin
155 μL
Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
FDA; OSHA; CDC
Videos
SDS
Documents

Documents

Promotions

Promotions