missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plastic Coplin Jar

Eisco™ Plastic Coplin Jar includes a screw cap cover and is designed to be used for staining and washing slides.

Supplier:  Eisco™ BI0108

Catalog No. S89498


Only null left

Description

Description

  • Polypropylene jar
  • Holds 5 slides (or 10 slides back to back)
  • Fits 25 x 75 mm slides
  • Threaded screw top

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Closure Included
Round
White
Polypropylene
Non-sterile
Staining Jar
1
6.35 cm
11.4 cm
Autoclavable
5 Slides
Reusable
Cylindrical
Non-treated
Staining slides
5
6.35 cm
6.35 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions