United Scientific™ Plastic Insulated Copper Wire, 100-Foot Roll
Useful in a wide variety of electricity demonstrations.
Supplier: United Scientific™ WCP22BK
Description
- 22-gauge (SWG) copper wire.
- Supplied in a 100-foot roll.
- Available with white, black, or red plastic insulation.
Specifications
Black | |
Wire | |
Copper |
Warning: Cancer and reproductive harm