missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plastic Pail with Tear Tab Lid

Durable HDPE pail with reusable tear tab lid.

$14.64 - $40.71

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Non-UN
Color White
For Use With (Application) Packaging and Shipping
Includes Tear tab Lid
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (English) Handle Material Price Quantity  
Catalog Number Mfr. No. Capacity (English) Handle Material Price Quantity  
19-058-150
View Documents
Fisherbrand™
2000FB
1 gal. 12 Gauge Galvanized Wire
Each for $14.64
Only null left
 
19-058-151
View Documents
Fisherbrand™
2100FB
2 gal. 10 Gauge Galvanized Wire
Each for $19.83
Only null left
 
19-058-152
View Documents
Fisherbrand™
2200FB
3.5 gal. 9 Gauge Galvanized Wire
Each for $27.96
Only null left
 
19-058-153
View Documents
Fisherbrand™
2300FB
5 gal. 9 Gauge Galvanized Wire
Each for $40.71
Only null left
 
Description

Description

  • Prime High Density Polyethylene (HDPE) construction
  • Round, open head pail
  • Reusable tear tab lid
  • Galvanized wire handle
  • White
Specifications

Specifications

Not Autoclavable
White
Tear tab Lid
Non-UN
Packaging and Shipping
1 Ea.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'