missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Plastic Weighing Scoops

Eisco™ Plastic Weighing Scoops are constructed of polypropylene and are the ideal replacement for metallic scoops.

Supplier:  Eisco™ CH0461FPK6

 View more versions of this product

Catalog No. S29436


Explore available promotions

Description

Description

  • Plastic weigh scoop with handle
  • Made from polypropylene
  • Autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Scoop
Autoclavable
100 mL
Measuring chemicals
12 cm
7 cm
Polypropylene
3.38 oz.
White
6
7 cm
SDS
Documents

Documents

Promotions

Promotions