Thermo Scientific™ Plate Count Agar
Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.
Supplier: Thermo Scientific™ RE09642
Description
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.
This standard medium meets the formulation of APHA, AOAC and PHLS.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
20mm x 150mm Tube | |
Plate Count Agar | |
20/Pk. |
United States | |
Agar |