missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

United Scientific™ Plate Glass Mirror Strips DFS Item

Mirror strips ideal for optics demonstrations.

Supplier:  United Scientific™ MSG4X4

Catalog No. S01317


Explore available promotions

Description

Description

  • Made from plate glass about 2 mm thick.
  • Highly silvered back.
Specifications

Specifications

4 x 4 in.
Mirror
2 mm
Glass (Silvered)
Square
Videos
SDS
Documents

Documents

Promotions

Promotions