Learn More
Platinum(II) chloride, 73% Pt, Thermo Scientific Chemicals
$328.13 - $1296.22
Chemical Identifiers
CAS | 10025-65-7 |
---|---|
Molecular Formula | Cl2Pt |
Molecular Weight (g/mol) | 265.98 |
MDL Number | MFCD00011181 |
InChI Key | CLSUSRZJUQMOHH-UHFFFAOYSA-L |
Synonym | Platinous chloride |
IUPAC Name | platinum(2+) dichloride |
SMILES | [Cl-].[Cl-].[Pt++] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC195380010
|
Thermo Scientific Chemicals
195380010 |
1 g | Glass bottle |
Each for $328.13
|
|
||||
AC195380050
|
Thermo Scientific Chemicals
195380050 |
5 g | Glass bottle |
Each for $1,296.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-65-7 | |
265.98 | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
platinum(2+) dichloride |
Cl2Pt | |
MFCD00011181 | |
Platinous chloride | |
[Cl-].[Cl-].[Pt++] |
Specifications
581.0°C | |
Brown to Gray-Green | |
1 g | |
PtCl2 | |
02,272 | |
Platinous chloride | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
platinum(2+) dichloride | |
266 | |
Glass bottle | |
6.0500g/mL |
10025-65-7 | |
Powder | |
Cl2Pt | |
MFCD00011181 | |
15,7643 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid, insoluble in alcohol and ether | |
[Cl-].[Cl-].[Pt++] | |
265.98 | |
73% (Pt) | |
6.05 | |
Platinum(II) chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 233-034-1
RUO – Research Use Only