missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ SuperFi II DNA Polymerase DFS Item

Invitrogen Platinum SuperFi II DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with an innovative buffer, enabling universal primer annealing for the highest success in PCR.

Supplier:  Invitrogen™ 12361010

 View more versions of this product

Catalog No. 12361010


Explore available promotions

Description

Description

Invitrogen Platinum SuperFi II DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with an innovative buffer, enabling universal primer annealing for the highest success in PCR. Due to the unique composition of the SuperFi II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules.

Features of Platinum SuperFi II DNA Polymerase include:

  • Exceptional >300X Taq fidelity
  • Universal primer annealing at 60°C
  • Superior specificity, sensitivity, and yields
  • Robust amplification of difficult-to-amplify targets

Platinum SuperFi II DNA Polymerase is an engineered enzyme with high processivity and increased resistance to PCR inhibitors. It also enables fast-cycling protocols and amplification of long targets (up to 20 kb). Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation. This technology also enables reaction setup at room temperature and provides increased sensitivity and yield.

Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize the annealing temperature for each primer pair. With Platinum SuperFi II DNA Polymerase, different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR
Specifications

Specifications

• Platinum SuperFi II DNA Polymerase (100 μL)
• 5X SuperFi II Buffer (1.25 mL)

Store at –20°C.
Platinum SuperFi II DNA Polymerase
100 reactions
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Blunt
20 kb or less
High
Fast
Dry Ice
>300X
100 Reactions
Standalone
Videos
SDS
Documents

Documents

Promotions

Promotions