missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum wire, 0.1mm (0.004in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

$227.47 - $847.90

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10959G1
View Documents
Thermo Scientific Chemicals
010959G1
1 m
Each for $227.47
 
AA10959G5
View Documents
Thermo Scientific Chemicals
010959G5
5 m
Each for $847.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

White
Silvery white wire
99.997% (Metals basis)
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Wire
7440-06-4
21.45g/cm3 at 20°C
Pt
1 m
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Odorless
Platinum
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only