Eisco™ Platinum Wire Loop
Housed in a glass tube and fused onto a glass handle for ease of use and safety. Eisco™ Platinum Wire Loop is designed for use in flame tests.
Supplier:  Eisco™ CH0538
Catalog No. S39825
Explore available promotions
Description
Description
  • Platinum wire loop
  • Supplied in borosilicate glass tube
  • Wire loop is fused onto a glass handle
Specifications
Specifications
Wire Loop
1
13.5 cm
Flame Test
Platinum
Product Suggestions
Product Suggestions