missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ PNPP Substrates

Thermo Scientific™ Pierce PNPP Substrates are powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications.

Supplier:  Thermo Scientific™ 37620

 View more versions of this product

Catalog No. PI37620


Explore available promotions

Description

Description

PNPP (p-Nitrophenyl Phosphate, Disodium Sal) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution, and a complete kit containing substrate solution and diethanolamine buffer.

Thermo Scientific Pierce PNPP (p-Nitrophenyl Phosphate, Disodium Salt) is a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm.

Features of PNPP (p-nitrophenyl phosphate):
• One-component alkaline phosphatase (AP) substrate
• Absorbance maximum of 405 nm
• Develops a yellow hue in solution upon reaction with AP
• Safer to use—contains a non-mercury, non-azide preservative

The substrate is offered in several formats:
PNPP Substrate
• Powder Form

PNPP Substrate Tablets
• 5 mg tablets with an inert filler as the balance

PNPP Substrate Kit
• PNPP tablets and a Diethanolamine Buffer to yield more than one liter of substrate. The Diethanolamine Substrate Buffer (Cat. No. 34064) is also provided individually as a 5X concentrate.

1-Step™ PNPP Substrate Solution
• Circumvents this time-consuming and variable introducing step by providing a single component PNPP substrate. This substrate is stable for 12 months at 2–8°C and can be stopped with conventional methods.

Specifications

Specifications

PNPP Substrate Kit
105 tablets kit
Pierce™
Alkaline Phosphatase
ELISA Substrate
Colorimetric
Chemical Substrate
Sufficient for: 5 mL of typical 1X substrate solution per tablet (525 mL total, approx. 5250 wells at 100 μL per well)
• Diethanolamine Buffer, 225 mL
• PNPP Tablets, 105 tablets (5 mg/tablet)

Store buffer at 4°C and PNPP tablets at -20°C.
Videos
SDS
Documents

Documents

Promotions

Promotions