Kova™ POC Urinalysis Dropper Controls
Urinalysis controls for smaller routine labs and point-of-care testing sites.
Supplier: Kova™ 88205
Description
- Designed for use in POC sites and smaller labs
- Control values assigned for the most commonly used dipsticks and instruments
- After opening, control is stabile for 30 days at room temp or until expiration when stored in refrigerator
- Barcoded labels (lot/expiration)
- Ready-to-use liquid controls in 2 levels (Normal/Abnormal)
- 22 month shelf life from date of manufacture
Specifications
CLSI and CLIA, ANSI Z400.5-2004 | |
2°C to 8°C | |
Ready-to-use, liquid products intended for use as a control for qualitative and semi-quantitative procedures used in routine urinalysis |
Yellow, green or amber | |
pH, Microalbumin, Glucose, Ketones, Protein, Hemoglobin, Ketones, Urobilinogen, Hemoglobin, Leukocytes, Urobilinogen, Specific Gravity, Leukocytes | |
2 x 5 mL |