missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polar Ware™ Stainless Steel Griffin Beakers
Ensure accurate measurement and safe transfer of liquids
$255.84 - $262.81
Specifications
Material | Stainless Steel |
---|---|
Label Type | Capacity-Full Mark Near Top |
Type | Beaker |
Style | Griffin |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
02-584-26E
|
Polar Ware™
VLL2000B |
2000 mL |
Each for $262.81
|
|
|||||
02-584-26H
|
Polar Ware™
VLL3000B |
3000 mL |
Each for $255.84
|
|
|||||
Description
- Made of stainless steel
- Available in different size capacities
Specifications
Stainless Steel | |
Capacity-Full Mark Near Top | |
Beaker | |
Griffin |