Cole-Parmer™ Essentials 100-Place Polypropylene Storage Boxes
Featuring reinforced lock and seal for secure storage
Supplier: Cole-Parmer™ Essentials 0675525
Description
- Reinforced triple hinges prevent cracks or breaks
- Bold alpha-numeric printing allows accurate tracking of samples
- Click fit lock ensures box will not open accidentally—large clasp allows easy opening with gloves
Specifications
Orange | |
Cryo Box | |
5 Pack | |
6.1 cm |
Polypropylene | |
0.5, 1.5 and 2 mL Tubes | |
2 in. |