MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Supplier:  MilliporeSigma™ CDUFBI0A1
 View more versions of this product
Catalog No. CDUFBI0A1
Explore available promotions
Description
Description
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q™ IQ 7000 system.
Specifications
Specifications
Delivered hermetically sealed
Biopak™ Polisher
No
Milli-Q IQ 7000 Ultrapure Water System
Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q IQ 7000 system