MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Supplier:  MilliporeSigma™ EDSPAK0A1
 View more versions of this product
Catalog No. EDSPAK0A1
Explore available promotions
Description
Description
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
  • Delivered hermetically sealed and with its protective bell.
  • To be rinsed with high grade methanol before use. 
  • Certificate of Quality available online
Specifications
Specifications
Delivered hermetically sealed
EDS-Pak™Â Polisher
No
Milli-Q IQ 7000 Ultrapure Water System
Production of endocrine disruptor-free water at the point of dispense of the Milli-Q IQ 7000 system