MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Supplier: MilliporeSigma™ LCPAK00A1
Description
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
- Delivered hermetically sealed and with its protective bell.
- To be rinsed with high grade methanol before use.
- Certificate of Quality available online.
Specifications
Delivered hermetically sealed | |
LC-Pak™ Polisher | |
No |
Milli-Q IQ 7000 Ultrapure Water System | |
Removal of trace organics for applications such as UHPLC, LC-MS or LC-MS/MS at the point of dispense of the Milli-QÂ IQ 7000 system |