missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poloxamer 188, Thermo Scientific Chemicals
$289.78 - $472.60
Chemical Identifiers
Molecular Formula | C5H10O2 |
---|---|
Synonym | Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ6608736
|
Thermo Scientific Chemicals
J6608736 |
500 g |
Each for $289.78
|
|
||||||
AAJ66087A1
|
Thermo Scientific Chemicals
J66087A1 |
1 kg |
Each for $472.60
|
|
||||||
Description
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPoloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
Solubility
Miscible with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
C5H10O2 | |
Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer |
Specifications
9003-11-6 | |
Ambient temperatures | |
C5H10O2 | |
Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer | |
Poloxamer 188 |
White | |
500 g,500 g | |
Miscible with water. | |
Solid |
RUO – Research Use Only