Poltex Countertop Serological Pipet Holder DFS Item
Holder keeps pipets tidy and convenient when used on countertop
Supplier:  Poltex RGSRPIPAC
Catalog No. 22000187
Explore available promotions
Description
Description
  • Shelves are removable for easy cleaning
  • Slanted top allows for extra storage of other items
  • Pocket dimensions:
    • 8.89cm (3.5 in.) wide
    • 27.94cm (11 in.) deep
    • 5.71, 6.98, 8.25, 10.79cm (2.25, 2.75, 3.25 and 4.25 in.) tall
Specifications
Specifications
Pipet Holder
4 Tiers
Serological Pipets
28.41 x 9.84 x 40.64cm (11.187 x 3.875 x 16 in.)