missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poltex Microtube Tube Rack Organizer Tray
Stack your 1.5, or 0.5ml microtube racks vertically to preserve precious benchtop space
Supplier: Poltex 5LGMICRO
Description
- Features 4 or 6 closed shelves
- Inside shelf Dimension: 2.25in. T×5in. W×8in. D
- Added storage space on top
- Sturdy acrylic
Specifications
Microtube Rack Tray | |
Acrylic | |
8 x 5 x 2.25 in. |
Racks | |
4 to 5 |