missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), 25 wt% solution in water; approx. M.W. 240,000

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol

Supplier:  Thermo Scientific Chemicals 185015000

 View more versions of this product

Catalog No. AC185015000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Poly(acrylic acid)
Liquid
100.0°C
[-CH2CH(CO2H)-]n
1.0900g/mL
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
prop-2-enoic acid
6581
23 to 27% (solids)
Glass bottle
25 wt. % Solution in Water
9003-01-4,7732-18-5
(C3H4O2)n
MFCD00084394
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
500 g
OC(=O)C(-*)C-*
72.06
CHEBI:18308
Authentic
1.09

Chemical Identifiers

9003-01-4
72.06
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
6581
OC(=O)C(-*)C-*
(C3H4O2)n
MFCD00084394
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
CHEBI:18308
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AT4680000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only