missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly-D-Lysine Hydrobromide, MP Biomedicals
$72.14 - $72.14
Chemical Identifiers
CAS | 27964-99-4 |
---|---|
Molecular Formula | C6H15BrN2O2 |
Molecular Weight (g/mol) | 227.102 |
InChI Key | MEXAGTSTSPYCEP-NUBCRITNSA-N |
Synonym | d-lysine hydrobromide |
PubChem CID | 87493163 |
IUPAC Name | (2R)-2,6-diaminohexanoic acid;hydrobromide |
SMILES | C(CCN)CC(C(=O)O)N.Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10269410
|
MP Biomedicals Inc
0210269410 |
10 mg |
Each for $72.14
|
|
|||||
Chemical Identifiers
27964-99-4 | |
227.102 | |
d-lysine hydrobromide | |
(2R)-2,6-diaminohexanoic acid;hydrobromide |
C6H15BrN2O2 | |
MEXAGTSTSPYCEP-NUBCRITNSA-N | |
87493163 | |
C(CCN)CC(C(=O)O)N.Br |
Specifications
27964-99-4 | |
d-lysine hydrobromide | |
MEXAGTSTSPYCEP-NUBCRITNSA-N | |
(2R)-2,6-diaminohexanoic acid;hydrobromide | |
87493163 | |
Poly-D-Lysine Hydrobromide |
C6H15BrN2O2 | |
10 mg | |
C(CCN)CC(C(=O)O)N.Br | |
227.102 | |
4,000 to 15,000 |
Safety and Handling
Recommended Storage : 0°C