missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(dimethylsiloxane), 100 cSt
CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
$181.62 - $271.76
Chemical Identifiers
| CAS | 9016-00-6 |
|---|---|
| Molecular Formula | (C2H6OSi)n |
| Molecular Weight (g/mol) | 74.15 |
| MDL Number | MFCD00084411 |
| InChI Key | SEUDSDUUJXTXSV-UHFFFAOYSA-N |
| Synonym | Simethicone, Dimethicone, PDMS |
| IUPAC Name | dimethylsilanone |
| SMILES | C[Si](C)(-*)O-* |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC178442500
|
Thermo Scientific Chemicals
178442500 |
250 g | Glass bottle |
Each for $181.62
|
|
||||
|
AC178445000
|
Thermo Scientific Chemicals
178445000 |
500 g | Glass bottle |
Each for $271.76
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9016-00-6 | |
| 74.15 | |
| SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
| dimethylsilanone |
| (C2H6OSi)n | |
| MFCD00084411 | |
| Simethicone, Dimethicone, PDMS | |
| C[Si](C)(-*)O-* |
Specifications
| Oil, Free From Suspended Matter | |
| (C2H6OSi)n | |
| MFCD00084411 | |
| 10, 8374 | |
| 250 g | |
| C[Si](C)(-*)O-* | |
| 74.15 | |
| Passes Test | |
| 1.4030 to 1.4050 (20°C, 589nm) | |
| Poly(dimethylsiloxane) |
| 9016-00-6 | |
| {Si(CH3)2O-}x | |
| 96%5g/mL | |
| Simethicone, Dimethicone, PDMS | |
| SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
| dimethylsilanone | |
| Authentic | |
| Glass bottle | |
| 100mPa·s (25°C) |
Safety and Handling
RTECSNumber : TQ2690000
RUO – Research Use Only