missing translation for 'onlineSavingsMsg'
Learn More

Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da

CAS: 26780-50-7 | (C3H4O2)x(C2H2O2)y | NaN g/mol

$272.31 - $272.31

Chemical Identifiers

CAS 26780-50-7
Molecular Formula (C3H4O2)x(C2H2O2)y
Molecular Weight (g/mol) NaN
MDL Number MFCD00131930
InChI Key LCSKNASZPVZHEG-UHFFFAOYNA-N
Synonym vicryl, poly d,l-lactide-co-glycolide, polyglactin 370 usan, polyglactin, 1,4-dioxane-2,5-dione-3,6-dimethyl-1,4-dioxane-2,5-dione 1:1, polyglactin 910 usan, vicryl rapide, vicryl plus
PubChem CID 71391
IUPAC Name 1,4-dioxane-2,5-dione; 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC(-*)C(=O)OCC(=O)O-*
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436200010
SDS View Documents
Thermo Scientific Chemicals
436200010
1 g Glass bottle
Each for $272.31
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26780-50-7
NaN
LCSKNASZPVZHEG-UHFFFAOYNA-N
71391
CC(-*)C(=O)OCC(=O)O-*
(C3H4O2)x(C2H2O2)y
MFCD00131930
vicryl, poly d,l-lactide-co-glycolide, polyglactin 370 usan, polyglactin, 1,4-dioxane-2,5-dione-3,6-dimethyl-1,4-dioxane-2,5-dione 1:1, polyglactin 910 usan, vicryl rapide, vicryl plus
1,4-dioxane-2,5-dione; 3,6-dimethyl-1,4-dioxane-2,5-dione

Specifications

Tan to White
26780-50-7
MFCD00131930
Solubility in water: soluble.
LCSKNASZPVZHEG-UHFFFAOYNA-N
1,4-dioxane-2,5-dione; 3,6-dimethyl-1,4-dioxane-2,5-dione
71391
0.15 to 0.25 dL/g
Crystalline Powder or Granules
(C3H4O2)x(C2H2O2)y
vicryl, poly d,l-lactide-co-glycolide, polyglactin 370 usan, polyglactin, 1,4-dioxane-2,5-dione-3,6-dimethyl-1,4-dioxane-2,5-dione 1:1, polyglactin 910 usan, vicryl rapide, vicryl plus
1 g
CC(-*)C(=O)OCC(=O)O-*
NaN
Glass bottle
Poly(DL-lactide-co-glycolide)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only