missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol), average M.W. 400
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
$166.69 - $782.35
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | PEG |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC192230010
|
Thermo Scientific Chemicals
192230010 |
1 L | Glass Bottle |
Each for $166.69
|
|
||||
|
AC192230051
|
Thermo Scientific Chemicals
192230051 |
5 L | Plastic drum |
Each for $782.35
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| PEG | |
| [H]OCCO |
Specifications
| Colorless | |
| 25322-68-3 | |
| 4.0°C to 8.0°C | |
| H(OCH2CH2)nOH | |
| 1.1275g/mL | |
| PEG | |
| 1 L | |
| [H]OCCO | |
| 62.07 | |
| 235°C | |
| Glass Bottle | |
| 1.1275 | |
| Poly(ethylene glycol) |
| Liquid | |
| >200.0°C | |
| (C2H4O)n | |
| MFCD01779601 | |
| 15, 7688 | |
| Solubility in water: soluble. Other solubilities: soluble in many organic solvents | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 400 | |
| 267 to 295mg KOH/g | |
| 1.4650 to 1.4680 | |
| 110-125 mPa.s (20°C) |
Safety and Handling
EINECSNumber : 500-038-2
RUO – Research Use Only