missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol methyl ether), average M.W. 500
CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
$208.91 - $208.91
Chemical Identifiers
| CAS | 9004-74-4 |
|---|---|
| Molecular Formula | (C2H4O)nCH4O |
| Molecular Weight (g/mol) | 76.10 |
| MDL Number | MFCD00084416 |
| InChI Key | XNWFRZJHXBZDAG-UHFFFAOYSA-N |
| Synonym | methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em |
| PubChem CID | 8019 |
| ChEBI | CHEBI:46790 |
| SMILES | COCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC387895000
|
Thermo Scientific Chemicals
387895000 |
500 g | Glass bottle |
Each for $208.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9004-74-4 | |
| 76.10 | |
| XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
| 8019 | |
| COCCO |
| (C2H4O)nCH4O | |
| MFCD00084416 | |
| methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
| CHEBI:46790 |
Specifications
| 9004-74-4 | |
| Colorless | |
| 4.5 to 7.5 (5% aq. soln.) | |
| Authentic | |
| (C2H4O)nCH4O | |
| MFCD00084416 | |
| 1.08 | |
| Solubility in water: soluble | |
| COCCO | |
| 76.10 | |
| 20 mm2/s (50°C) | |
| Liquid |
| 12°C | |
| 1.0800g/mL | |
| 268°C | |
| Glass bottle | |
| CH3(OCH2CH)nOH | |
| 500 g | |
| methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
| XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
| 2-methoxyethanol | |
| 8019 | |
| CHEBI:46790 | |
| Poly(ethylene glycol methyl ether) |
RUO – Research Use Only