missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol methyl ether), average M.W. 350
CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
$239.74 - $239.74
Chemical Identifiers
| CAS | 9004-74-4 |
|---|---|
| Molecular Formula | (C2H4O)nCH4O |
| Molecular Weight (g/mol) | 76.10 |
| MDL Number | MFCD00084416 |
| InChI Key | XNWFRZJHXBZDAG-UHFFFAOYSA-N |
| Synonym | methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em |
| PubChem CID | 8019 |
| ChEBI | CHEBI:46790 |
| IUPAC Name | 2-methoxyethanol |
| SMILES | COCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC192305000
|
Thermo Scientific Chemicals
192305000 |
500 g | Glass bottle |
Each for $239.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9004-74-4 | |
| 76.10 | |
| XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
| 8019 | |
| 2-methoxyethanol |
| (C2H4O)nCH4O | |
| MFCD00084416 | |
| methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
| CHEBI:46790 | |
| COCCO |
Specifications
| 4.0 to 8.0 (1% aq. soln.) | |
| 9004-74-4 | |
| CH3(OCH2CH)nOH | |
| 1.09g/mL | |
| 500 g | |
| COCCO | |
| 76.10 | |
| CHEBI:46790 | |
| Authentic | |
| 1.4560 to 1.4580 (20°C, 589nm) | |
| Poly(ethylene glycol methyleether), Average M.W. 350 |
| Liquid | |
| (C2H4O)nCH4O | |
| MFCD00084416 | |
| methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
| XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
| 2-methoxyethanol | |
| 8019 | |
| 152 to 168mg KOH/g | |
| Glass bottle | |
| 0.15% max. (K.F.) |
Safety and Handling
RTECSNumber : PE8263000
TSCA : TSCA
RUO – Research Use Only