missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(methyl methacrylate), 600 micron, Thermo Scientific Chemicals

$180.62 - $597.16

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4396918
View Documents
Thermo Scientific Chemicals
04396918
50 g
Each for $180.62
 
AA4396930
View Documents
Thermo Scientific Chemicals
04396930
250 g
Each for $597.16
 
Description

Description

Poly(methyl methacrylate), 600 microns is used in many medical and biochemical applications. It is suitable for 2D and 3D particle assemblies and for investigations in the colloidal crystal field due to the high monodispersity. It is also used in the field of one- and two-component toners for copying machines. Further, it is having a reduced non-specific protein binding activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9011-14-7
NaN
PMAMJWJDBDSDHV-UHFFFAOYSA-N
COC(=O)C(C)(-*)C-*
(C5H8O2)n
MFCD00134349
1-methoxy-2-methyl-1-oxopropan-2-yl

Specifications

Powder
(C5H8O2)n
Soluble in tetrahydrofuran,toluene,cyclohexanone,ethyl acetate and chloroform. Insoluble in water,alcohols and aliphatic hydrocarbons.
PMAMJWJDBDSDHV-UHFFFAOYSA-N
1-methoxy-2-methyl-1-oxopropan-2-yl
600 Micron
9011-14-7
MFCD00134349
50 g
COC(=O)C(C)(-*)C-*
NaN
Poly (methyl methacrylate)
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only