missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(methyl Vinyl Ether) (30% in Water), TCI America™
$63.05 - $192.44
Chemical Identifiers
CAS | 9003-09-2 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00161528 |
InChI Key | XJRBAMWJDBPFIM-UHFFFAOYSA-N |
Synonym | Methyl Vinyl Ether Polymer, Poly(vinyl Methyl Ether), Vinyl Methyl Ether Polymer |
PubChem CID | 7861 |
IUPAC Name | methoxyethene |
SMILES | COC=C |
Chemical Identifiers
9003-09-2 | |
58.08 | |
XJRBAMWJDBPFIM-UHFFFAOYSA-N | |
7861 | |
COC=C |
C3H6O | |
MFCD00161528 | |
Methyl Vinyl Ether Polymer, Poly(vinyl Methyl Ether), Vinyl Methyl Ether Polymer | |
methoxyethene |
Specifications
9003-09-2 | |
MFCD00161528 | |
Methyl Vinyl Ether Polymer, Poly(vinyl Methyl Ether), Vinyl Methyl Ether Polymer | |
COC=C | |
58.08 | |
Liquid |
C3H6O | |
25 g | |
XJRBAMWJDBPFIM-UHFFFAOYSA-N | |
methoxyethene | |
7861 | |
Poly(methyl Vinyl Ether) (30% in Water) |
Safety and Handling
EINECSNumber : (6)-0982
TSCA : Yes