Learn More
Poly(propylene glycol) bis(2-aminopropyl ether), Mn ca 230, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H37543AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly(propylene glycol) bis(2-aminopropyl ether) | |
9046-10-0 | |
0.948 g/mL | |
100 mL | |
3,4-pyrrolidinediol, 3,4-dihydroxy-2h-pyrroline, polyetheramine d 230, polyetheramine d 2000, poly propylene glycol bis 2-aminopropyl ether, poly propylene oxide , diamine terminated, o,o'-bis 2-aminopropyl polypropyleneglycol, o,o'-bis 2-aminopropyl polypropylene glycol 300, polypropylene glycol 130 bis 2-aminopropyl ether, polypropylene glycol 300 bis 2-aminopropyl ether | |
OC1CNCC1O | |
103.12 | |
Liquid |
Mn ≈230 | |
Colorless to Yellow | |
C4H9NO2 | |
UN2922 | |
JCZPOYAMKJFOLA-UHFFFAOYNA-N | |
pyrrolidine-3,4-diol | |
13329881 |
Chemical Identifiers
9046-10-0 | |
103.12 | |
3,4-pyrrolidinediol, 3,4-dihydroxy-2h-pyrroline, polyetheramine d 230, polyetheramine d 2000, poly propylene glycol bis 2-aminopropyl ether, poly propylene oxide , diamine terminated, o,o'-bis 2-aminopropyl polypropyleneglycol, o,o'-bis 2-aminopropyl polypropylene glycol 300, polypropylene glycol 130 bis 2-aminopropyl ether, polypropylene glycol 300 bis 2-aminopropyl ether | |
pyrrolidine-3,4-diol |
C4H9NO2 | |
JCZPOYAMKJFOLA-UHFFFAOYNA-N | |
13329881 | |
OC1CNCC1O |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECSNumber : TR3702500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only