Corning™ Polycarbonate Erlenmeyer Flask with Dip Tube, Male Luer Lock, Sterile
Provide optimum visibility and durability
Supplier:  Corning™ 11415
 View more versions of this product
Catalog No. 15-100-151
Explore available promotions
Description
Description
Corning™ closed system Erlenmeyer flasks will enhance the productivity of your shaker culture operations when adding or removing medium and cells. Flask is made from optically clear polycarbonate and is preassembled with an aseptic transfer cap. Unit is sterilized by gamma radiation and is nonpyrogenic and RNase-/DNase-free.
Flask:
  • Polycarbonate construction: USP Class VI material
  • Molded-in graduations for accuracy (+/-5%)
Aseptic Transfer Cap:
  • Chemically resistant, heat-sealable, heat-sealable, flexible tubing, 1/8” ID, 1/4” OD
  • Male Luer Lok Disconnect
Specifications
Specifications
8.4 oz.
Polypropylene
38 mm
Polycarbonate
Single-use
Gamma-irradiated
Orange
Erlenmeyer Flask with 1/8 in. Tube
250 mL
0.2μm Vent, 1/8 in. dip tube, male luer lock
Threaded
Aseptic Transfer Cap
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.