missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PolyCo™ VR™ Grommet Aprons
Blue aprons feature seamless durability and strength. Ideal for critical and controlled environments.
Supplier: PolyCo™ 45420
Description
- Constructed of proprietary PolyCo™ VR™ material from top to bottom, including the grommets, for an environmentally safe, more durable product
- Comfortable, adjustable ties for neck and waist allow for flexible positioning and custom fit
- Strong, texturized nylon ties designed for extended wear; interchangeable for better sanitation
Specifications
Controlled Environments Apron | |
Blue | |
Ties | |
8 mil | |
90 x 114 cm (35 x 45 in.) |
VR™ Proprietary Film | |
Reusable | |
Non-sterile | |
Critical and Controlled Environments |