missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polydimethylsiloxane, hydroxy terminated, M.W. 4200, Thermo Scientific Chemicals

$50.58 - $773.15

Chemical Identifiers

CAS 70131-67-8
Molecular Formula HOSi
Molecular Weight (g/mol) 45.09
MDL Number MFCD01325012
InChI Key XQSFXFQDJCDXDT-UHFFFAOYSA-N
Synonym silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
PubChem CID 6327522
IUPAC Name hydroxysilicon
SMILES O[Si]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43769AD
View Documents
Thermo Scientific Chemicals
043769AD
50 mL
Each for $50.58
 
AA43769AK
View Documents
Thermo Scientific Chemicals
043769AK
250 mL
Each for $109.97
 
AA43769K2
View Documents
Thermo Scientific Chemicals
043769K2
1 L
Each for $335.41
 
AA43769K4
View Documents
Thermo Scientific Chemicals
043769K4
2.5 L
Each for $773.15
 
Description

Description

Polydimethylsiloxane is frequently used in a liquid containing 4-5% silica gel, emulsifiers and preservatives. It is employed in applications like contact lenses, medical devices, elastomers, in shampoos, food (antifoaming agent), caulking, lubricants, kinetic sand , and heat-resistant tiles. PDMS is a common surfactant, component of defoamers and as an herbicidal penetran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70131-67-8
45.09
XQSFXFQDJCDXDT-UHFFFAOYSA-N
6327522
O[Si]
HOSi
MFCD01325012
silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
hydroxysilicon

Specifications

Liquid
HOSi
0.97 g/mL
Soluble in carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Insoluble in water and ethanol.
XQSFXFQDJCDXDT-UHFFFAOYSA-N
hydroxysilicon
6327522
70131-67-8
MFCD01325012
silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
50 mL
O[Si]
45.09
Polydimethylsiloxane, hydroxy terminated, M.W. 4,200
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only