missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polydimethylsiloxane, Silanol Terminated, Spectrum™ Chemical
P2952, 70131-67-8
$250.97 - $1889.17
Chemical Identifiers
CAS | 70131-67-8 |
---|---|
Molecular Formula | HOSi |
Molecular Weight (g/mol) | 45.09 |
MDL Number | MFCD01325012 |
InChI Key | XQSFXFQDJCDXDT-UHFFFAOYSA-N |
IUPAC Name | hydroxy-λ¹-silanylidyne |
SMILES | O[Si] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18605294
|
Spectrum Chemical Mfg Cor
P2952100GM |
100 g | Amber Glass Bottle |
Each for $250.97
|
|
||||
18605295
|
Spectrum Chemical Mfg Cor
P29521KG |
1 kg | Amber Glass Bottle |
Each for $827.26
|
|
||||
18605296
|
Spectrum Chemical Mfg Cor
P29523KG |
3 kg | Amber Glass Bottle |
Each for $1,889.17
|
|
||||
Description
Spectrum™ Chemical Polydimethylsiloxane, Silanol Terminated is a silicon-based organic polymer with an average molecular weight of 70,000 - 80,000. It is used in cosmetic and personal care products as a surfactant and conditioning agent. It appears as a clear, viscous liquid and is insoluble in water.Chemical Identifiers
70131-67-8 | |
45.09 | |
XQSFXFQDJCDXDT-UHFFFAOYSA-N | |
O[Si] |
HOSi | |
MFCD01325012 | |
hydroxy-λ¹-silanylidyne |
Specifications
70131-67-8 | |
Amber Glass Bottle | |
MFCD01325012 | |
XQSFXFQDJCDXDT-UHFFFAOYSA-N | |
hydroxy-λ¹-silanylidyne | |
15000-20000 cSt |
1 | |
HOSi | |
100 g | |
O[Si] | |
45.09 | |
Reagent |