missing translation for 'onlineSavingsMsg'
Learn More

Polyester acrylate/methacrylate, PEAM-1769

Recommended for use as a base resin or an additive to reduce stress | CAS: 921214-61-1

$164.70 - $513.70

Chemical Identifiers

CAS 921214-61-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4635114
View Documents
Thermo Scientific Chemicals
04635114
25 g
Each for $164.70
Only null left
 
AA4635122
View Documents
Thermo Scientific Chemicals
04635122
100 g
Each for $513.70
Only null left
 
Description

Description

Recommended for use as a base resin or an additive to reduce stress

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

921214-61-1

Specifications

Amber
921214-61-1
Polyester acrylate/methacrylate, PEAM-1769
Amber liquid
25 g
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P272-P280g-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P333+P313-P362-P501c

H315-H317-H318-H335-H361

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only