Polyethylene Glycol 1450, NF, Spectrum™ Chemical
P1492, 25322-68-3, H(OCH2CH2)nOH
Supplier:  Spectrum Chemical Mfg Cor P149212KGBL
 View more versions of this product
Catalog No. 18605305
Explore available promotions
Description
Description
Spectrum™ Chemical Polyethylene Glycol 1450, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
4.5 to 7.5
1
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
NF
Poly Pail
25322-68-3
(C2H4O)n
12 kg
[H]OCCO
62.07
0.001
25-32 cSt
Chemical Identifiers
25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
[H]OCCO
(C2H4O)n
MFCD01779601
ethane-1,2-diol