missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyethylene Glycol 1500, NF, Spectrum™ Chemical

PO120, 25322-68-3, H(OCH2CH2)nOH

$257.00 - $2507.59

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605307
View Documents Promotion Details
Spectrum Chemical Mfg Cor
PO120500GM
500 g Poly Bottle
Each for $257.00
 
18605309
View Documents Promotion Details
Spectrum Chemical Mfg Cor
PO12012KG
12 kg Poly Pail
Each for $2,507.59
 
Description

Description

Spectrum™ Chemical Polyethylene Glycol 1500, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
[H]OCCO
(C2H4O)n
MFCD01779601
ethane-1,2-diol

Specifications

4.5 to 7.5
1
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
NF
Poly Bottle
25322-68-3
(C2H4O)n
500 g
[H]OCCO
62.07
0.001
26-33 cSt
SDS
missing translation for 'documents'

missing translation for 'documents'